Learn More
CD31/PECAM-1 Antibody (CL14693) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26877B
Product Code. 30290591
  • £558.00 / 0.10mL
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD31/PECAM-1
Monoclonal
Biotin
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
This antibody was developed using a synthetic peptide derived from P16284, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers
5175
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry
CL14693
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified