missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31/PECAM-1 Antibody (CL14693) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26877AF750
This item is not returnable.
View return policy
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Immunohistochemistry
Specifications
| CD31/PECAM-1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| This antibody was developed using a synthetic peptide derived from P16284, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
| 5175 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry | |
| CL14693 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction