missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31/PECAM-1 Antibody (C31/8242R), Novus Biologicals™
Rabbit Monoclonal Antibody
£232.00 - £412.00
Specifications
| Antigen | CD31/PECAM-1 |
|---|---|
| Clone | C31/8242R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636914
|
Novus Biologicals
NBP3-20291-100ug |
100 μg |
£412.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620295
|
Novus Biologicals
NBP3-20291-20ug |
20 μg |
£232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| CD31/PECAM-1 | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
| 10mM PBS with 0.05% BSA | |
| 5175 | |
| IgG κ | |
| Protein A or G purified |
| C31/8242R | |
| Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Recombinant fragment (aa 28-330) of human CD31/PECAM-1 expressed in E. Coli (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title