Learn More
Invitrogen™ CD31 Monoclonal Antibody (2H8)

Armenian Hamster Monoclonal Antibody
Brand: Invitrogen™ MA3105
Description
MA3105 targets CD31 in FACS, and Neu applications and shows reactivity with mouse and Human samples. The MA3105 immunogen is mouse PECAM-1 (CD31). MA3105 detects CD31 which has a predicted molecular weight of approximately 79 kDa. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.
Specifications
| CD31 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P16284, Q08481 | |
| Pecam1 | |
| Mouse PECAM-1 (CD31). | |
| 500 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Neutralization | |
| 2H8 | |
| Unconjugated | |
| Pecam1 | |
| C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
| Armenian Hamster | |
| Protein G | |
| RUO | |
| 18613, 5175 | |
| -20°C | |
| Liquid | |
| Responsibly packaged |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.