missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD305 (LAIR1) Monoclonal Antibody (NKTA255), NovaFluor™ Red 700, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£151.00 - £336.00

Specifications

Antigen CD305 (LAIR1)
Clone NKTA255
Concentration 0.4 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089359
View Documents
Invitrogen™
H081T02R03-A
25 Tests
£151.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30087454
View Documents
Invitrogen™
H081T03R03-A
100 Tests
£336.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD305 (LAIR1) Monoclonal antibody specifically detects CD305 (LAIR1) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD305 (LAIR1)
0.4 μg/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; Lair-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1
IgG1
Affinity chromatography
Antibody
NKTA255
Flow Cytometry
NovaFluor Red 700
Mouse
Human
Q6GTX8
3903
Primary
4°C, store in dark, DO NOT FREEZE!
LAIR1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.