missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD305 (LAIR1) Monoclonal Antibody (NKTA255), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£151.00 - £336.00
Specifications
| Antigen | CD305 (LAIR1) |
|---|---|
| Clone | NKTA255 |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088434
|
Invitrogen™
H081T02B08-A |
25 Tests |
£151.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089387
|
Invitrogen™
H081T03B08-A |
100 Tests |
£336.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD305 (LAIR1) Monoclonal antibody specifically detects CD305 (LAIR1) in Human samples. It is validated for Flow CytometrySpecifications
| CD305 (LAIR1) | |
| 0.8 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; Lair-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1 | |
| IgG1 | |
| Affinity chromatography | |
| Antibody |
| NKTA255 | |
| Flow Cytometry | |
| NovaFluor Blue 660-120S | |
| Mouse | |
| Human | |
| Q6GTX8 | |
| 3903 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| LAIR1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title