Learn More
CD303a, eFluor 450, clone: 201A, eBioscience™
Mouse Monoclonal Antibody
Brand: Invitrogen 48-9818-41
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.
Description: This 201A monoclonal antibody reacts with human CD303a, which is also known as CLEC4C or Blood Dendritic Cell Antigen 2 (BDCA-2). This type II transmembrane protein is expressed almost exclusively on plasmacytoid dendritic cells, which secrete type I interferon during antiviral and antitumor responses. Studies have demonstrated that CD303a binds asialo-galactosyl-oligosaccharides on invading pathogens, resulting in the activation of signaling pathways that lead to CD303a internalization and downregulation of interferon production.Applications Reported: This 201A antibody has been reported for use in flow cytometric analysis.Applications Tested: This 201A antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications
| CD303a | |
| Monoclonal | |
| 5μl/Test | |
| PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| Q8WTT0 | |
| CLEC4C | |
| Affinity chromatography | |
| RUO | |
| 170482 | |
| Store at 2-8°C. Do not freeze. Light-sensitive material. | |
| Liquid |
| Flow Cytometry | |
| 201A | |
| eFluor 450 | |
| CLEC4C | |
| C-type lectin superfamily member 7, Blood dendritic cell antigen 2 protein, BDCA-2, Dendritic lectin, CD303 antigen, CLEC4C, BDCA2, CLECSF11, CLECSF7, DLEC, HECL, C-type lectin domain family 4 member C | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.