missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD302 Mouse anti-Human, Mouse, Clone: 2B1E8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CD302 |
|---|---|
| Clone | 2B1E8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16868705
|
Proteintech
66640-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16858705
|
Proteintech
66640-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD302 is a C-type lectin receptor involved in cell adhesion and migration, as well as endocytosis and phagocytosis.Specifications
| CD302 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BIMLEC, C type lectin BIMLEC, CD302, CD302 antigen, CD302 molecule, CLEC13A, DCL 1, DCL1, KIAA0022 | |
| Cd302 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2B1E8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| Q8IX05, Q9DCG2 | |
| 66205, 9936 | |
| CD302 Fusion Protein Ag9873 | |
| Primary | |
| -20°C | |
| CD302 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title