missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD302/CLEC13A Rabbit anti-Mouse, Clone: 71, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90687-50UL
This item is not returnable.
View return policy
Description
CD302/CLEC13A Monoclonal specifically detects CD302/CLEC13A in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD302/CLEC13A | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 9936 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 71 | |
| ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 | |
| BIMLEC, CD302 antigenC-type lectin domain family 13, member A, CD302 molecule, CLEC13AMGC22301, C-type lectin domain family 13 member A, DCL-1, DCL1C-type lectin BIMLEC, KIAA0022FLJ43091, Type I transmembrane C-type lectin receptor DCL-1 | |
| Recombinant Mouse CD302/CLEC13A protein | |
| 50 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction