missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300LG/Nepmucin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98158-50ul
This item is not returnable.
View return policy
Description
CD300LG/Nepmucin Polyclonal antibody specifically detects CD300LG/Nepmucin in Rat samples. It is validated for ELISA
Specifications
| CD300LG/Nepmucin | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| CD300 antigen like family member G, CD300 antigen-like family member G, CD300 molecule like family member g, CD300 molecule-like family member g, CD300g antigen, CLM-9, CLM9Trem4, CMRF35-like molecule 9, NEPMUCIN, TREM4, TREM-4, Triggering receptor expressed on myeloid cells 4 | |
| Produced in rabbits immunized with purified, recombinant Rat CD300LG/Nepmucin (Accession#: XP_002724611.1; Met1-Arg160). | |
| 50 μg | |
| Primary | |
| Rat | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 146894 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction