missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD300f Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553080
This item is not returnable.
View return policy
Description
Immunogen sequence: SIVTQITGPT TVNGLERGSL TVQCVYRSGW EAYLKWWCRG AIWRDCKILV KTSGSEQEVK RDRVSIKDNQ KNRTFTVTME DLMKTDADTY WCGIEKTGND LGVTVQVTID PASTTASTPA PTTPTSTTFT APITQEETSS S Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 53%.
CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6; MIM 176883) or SHIP.
Specifications
| CD300f | |
| Polyclonal | |
| Unconjugated | |
| CD300LF | |
| CD300 antigen like family member F; CD300 antigen-like family member F; CD300 molecule like family member f; CD300 molecule-like family member f; CD300f; Cd300lf; CLIM1; CLM1; CLM-1; CMRF-35-like molecule 1; CMRF35-like molecule 1; DC-derived Ig-like receptor 2; Digr2; F730004D16Rik; IgSF13; Immune receptor expressed on myeloid cells 1; immunoglobin superfamily member 13; immunoglobulin superfamily member 13; inhibitory receptor IREM1; IREM1; IREM-1; Leukocyte mono-Ig-like receptor 3; Lmir3; MAIR-5; MAIR-V; myeloid-associated immunoglobulin-like receptor 5; NK inhibitory receptor; NKIR; Pigr3; UNQ3105/PRO10111 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 146722 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TDQ1 | |
| CD300LF | |
| Recombinant protein corresponding to Human CD300f. Recombinant protein control fragment (Product #RP-90561). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction