missing translation for 'onlineSavingsMsg'
Learn More

CD300e/LMIR6 Antibody (UP-H2), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18406985 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406985 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406985 Supplier Novus Biologicals Supplier No. NBP262230AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD300e/LMIR6 Monoclonal antibody specifically detects CD300e/LMIR6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300e/LMIR6
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone UP-H2
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CD300 antigen like family member E, CD300 antigen-like family member E, CD300e antigenPIgR-2, CD300e molecule, CD300LEPIgR2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM2IREM-2, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2
Host Species Mouse
Immunogen CD300e-HA-transfected cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 342510
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.