missing translation for 'onlineSavingsMsg'
Learn More

CD300c Antibody (105), Janelia Fluor™ 549 , Novus Biologicals™

Product Code. 18426429 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426429 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426429 Supplier Novus Biologicals Supplier No. NBP289993JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD300c Monoclonal antibody specifically detects CD300c in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300c
Applications ELISA
Classification Monoclonal
Clone 105
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CD300c antigenCMRF35 antigen, CD300c molecule, CLM-6, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IgSF16, IGSF16CD300 antigen-like family member C, Immunoglobulin superfamily member 16, LIR
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD300c (NP_006669.1; Met 1-Arg 183).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10871
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.