missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD300b Polyclonal Antibody
GREENER_CHOICE

Product Code. 15896873
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15896873 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15896873 Supplier Invitrogen™ Supplier No. PA568464

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with Human samples.

CD300 molecule-like family member b (CD300LB), also known as CLM-7, is a gene located on chromosome 17q25.1, encoding an immunoreceptor that belongs to the CD300 family of molecules. CD300LB is primarily expressed on the surface of myeloid cells, including dendritic cells and macrophages, where it plays an essential role in modulating immune responses. The receptor is involved in the regulation of cell activation and inflammatory responses through its interactions with phosphatidylserine and other lipid ligands, which can lead to either activation or inhibition of immune cell functions. CD300LB influences processes like antigen presentation, phagocytosis, and cytokine production, thereby impacting innate immune responses and the maintenance of immune homeostasis. Its expression and activity are of particular interest in contexts like autoimmunity, infection, and cancer, where it may contribute to disease progression and serve as a potential therapeutic target in modulating immune functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300b
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CD300LB
Gene Accession No. A8K4G0
Gene Alias CD300 antigen like family member B; CD300 antigen-like family member B; CD300 molecule like family member b; CD300 molecule-like family member b; CD300b; CD300LB; CLM7; CLM-7; CMRF35A2; CMRF35-A2; CMRF35-like molecule 7; CMRF-35-like molecule-7; Gm253; Immune receptor expressed on myeloid cells 3; IREM3; IREM-3; Leukocyte mono-Ig-like receptor 5; LMIR5; TREM5; TREM-5; triggering receptor expressed on myeloid cells 5; UNQ2530/PRO6029
Gene Symbols CD300LB
Host Species Rabbit
Immunogen Synthesized peptide derived from human CD300b [aa 61-110].
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124599
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.