missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD300a Monoclonal Antibody (MEM-260)

Description
This antibody reacts with an extracellular epitope of CD300a, a 60 kDa leukocyte transmembrane glycoprotein expressed on human granulocytes, monocytes, neutrophils, NK cells, mast cells and dendritic cells, 25% of circulating T cells and 15% of circulating B cells. Western Blot: non-reducing conditions.
The CMRF35 antigen (CMRF35A; MIM 606786), which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes. CMRF35H is recognized by the same antibody and is distinct from CMRF35 (Green et al., 1998 [PubMed 9701027]).
Specifications
Specifications
| Antigen | CD300a |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | MEM-260 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | CD300A |
| Gene Accession No. | Q9UGN4 |
| Gene Alias | B230315M08Rik; CD300 antigen-like family member A; Cd300a; CD300a antigen; CD300a molecule; Clm8; CLM-8; CMRF35H; CMRF-35H; CMRF35-H; CMRF35H leukocyte immunoglobulin-like receptor; CMRF35H9; CMRF-35-H9; CMRF35-H9; CMRF35HIRp60; CMRF35-like molecule 8; HSPC083; IGSF12; Immunoglobulin superfamily member 12; inhibitory receptor protein 60; IRC1; IRC1/IRC2; IRC2; IRp60; leukocyte membrane antigen; leukocyte mono-Ig-like receptor 1; Lmir1; MAIR-1; MAIR-I; MAIR-Ia; Mast cell-derived paired immunoglobulin-like receptor 1; Mcipr1; mcpir1; MMAC8; myeloid-associated immunoglobulin-like receptor 1; NK inhibitory receptor; NKRL; Pigr4; polymeric immunoglobulin receptor 4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?