missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300a/LMIR1 Antibody (MEM-260), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-58726PE
This item is not returnable.
View return policy
Description
CD300a/LMIR1 Monoclonal specifically detects CD300a/LMIR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD300a/LMIR1 | |
| Monoclonal | |
| PE | |
| Q9UGN4 | |
| CD300A | |
| HPB acute lymphoid leukemia cell line (NM_007261.2). | |
| 0.1 mL | |
| Mast Cell Markers | |
| 11314 | |
| Human | |
| Purified |
| Flow Cytometry | |
| MEM-260 | |
| Flow Cytometry | |
| CD300a antigenCMRF35H9, CD300a molecule, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, Irp60, leukocyte membrane antigen | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody (clone MEM-260) reacts with CD300a, a 60 kDa leukocyte transmembrane glycoprotein expressed on human granulocytes, monocytes, neutrophils, NK cells, mast cells and dendritic cells, 25% of circulating T cells and 15% of circulating B cells. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction