Learn More
CD300a/LMIR1 Antibody (172224), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB1186PECY55
Product Code. 30026807
  • £519.00 / 0.10mL
Description
CD300a/LMIR1 Monoclonal antibody specifically detects CD300a/LMIR1 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD300a/LMIR1
Monoclonal
PE-Cyanine5.5
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry
172224
Flow Cytometry
CD300a antigenCMRF35H9, CD300a molecule, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, Irp60, leukocyte membrane antigen
Mouse myeloma cell line NS0-derived recombinant mouse CD300a/LMIR1, Leu28-Arg183, Accession # BAC80268
0.1 mL
Mast Cell Markers
11314
Store at 4C in the dark. Do not freeze.
IgG2a