missing translation for 'onlineSavingsMsg'
Learn More

CD30/TNFRSF8 Antibody (Ki-1/1505R), DyLight 594, Novus Biologicals™

Product Code. 18416503 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416503 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416503 Supplier Novus Biologicals Supplier No. NBP254557DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD30/TNFRSF8 Monoclonal antibody specifically detects CD30/TNFRSF8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD30/TNFRSF8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone Ki-1/1505R
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Host Species Rabbit
Immunogen Recombinant human full-length CD30/TNFRSF8 protein (Uniprot: P28908)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 943
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.