Learn More
CD30/TNFRSF8 Antibody (CL12632) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18541JF669
Product Code. 30297058
  • £582.00 / 0.10mL
Description
CD30/TNFRSF8 Monoclonal antibody specifically detects CD30/TNFRSF8 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD30/TNFRSF8
Monoclonal
Janelia Fluor 669
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
This antibody was developed using a synthetic peptide derived from P28908, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Immunology, Stem Cell Markers
943
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12632
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified