missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30 Monoclonal Antibody (Ber-H2), Super Bright™ 702, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 67-0309-41
This item is not returnable.
View return policy
Description
Mouse Monoclonal Antibody
CD30 Monoclonal antibody specifically detects CD30 in Human samples. It is validated for Flow Cytometry
Specifications
| CD30 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P28908 | |
| TNFRSF8 | |
| Affinity chromatography | |
| RUO | |
| 943 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| Ber-H2 | |
| Super Bright 702 | |
| TNFRSF8 | |
| Ber-H2; Cd30; CD30 antigen; CD30 molecule; CD30L receptor; D1S166E; Ki; Ki-1; Ki-1 antigen; Lymphocyte activation antigen CD30; sCD30; soluble CD30; Tnfrsf8; Tumor necrosis factor receptor superfamily member 8 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction