missing translation for 'onlineSavingsMsg'
Learn More

CD30 Ligand/TNFSF8 Rat anti-Mouse, Clone: RM153, Novus Biologicals™

Product Code. 18363883 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363883 0.025 mg 25µg
18342813 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18363883 Supplier Novus Biologicals Supplier No. NBP3119920.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CD30 Ligand/TNFSF8 Monoclonal antibody specifically detects CD30 Ligand/TNFSF8 in Mouse samples. It is validated for Flow Cytometry, Neutralization, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD30 Ligand/TNFSF8
Applications Flow Cytometry, Neutralization, Functional Assay
Classification Monoclonal
Clone RM153
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Block/Neutralize, Functional Assay
Formulation PBS
Gene Alias CD153, CD153 antigen, CD30 antigen ligand, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8
Host Species Rat
Immunogen This antibody was raised by immunising Sprague Dawley rats with the CHO cells stably transfected with murine CD30 Ligand/TNFSF8 and fusing the splenocytes with P3U1 myeloma cells.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 944
Target Species Mouse
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.