missing translation for 'onlineSavingsMsg'
Learn More

CD30 Ligand/TNFSF8 Antibody (RM153), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18389901 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389901 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389901 Supplier Novus Biologicals Supplier No. NBP311991AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD30 Ligand/TNFSF8 Monoclonal antibody specifically detects CD30 Ligand/TNFSF8 in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD30 Ligand/TNFSF8
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone RM153
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD153, CD153 antigen, CD30 antigen ligand, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8
Host Species Rabbit
Immunogen This antibody was raised by immunising Sprague Dawley rats with the CHO cells stably transfected with murine CD30 Ligand/TNFSF8 and fusing the splenocytes with P3U1 myeloma cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 944
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.