missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 zeta Antibody (H146-968), CoraFluor™ 1, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-44683CL1
This item is not returnable.
View return policy
Description
CD3 zeta Monoclonal antibody specifically detects CD3 zeta in Human,Mouse samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Flow (Intracellular),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| CD3 zeta | |
| Monoclonal | |
| CoraFluor 1 | |
| CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA | |
| Synthetic peptide corresponding to amino acids 151-164 of mouse CD3 zeta. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 919 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 κ |
| Flow Cytometry, Immunoprecipitation, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| H146-968 | |
| PBS | |
| Armenian Hamster | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction