missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 zeta Antibody (21H6), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | CD3 zeta |
|---|---|
| Clone | 21H6 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043564
|
Novus Biologicals
NBP3-26684-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043528
|
Novus Biologicals
NBP3-26684-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 zeta Monoclonal antibody specifically detects CD3 zeta in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| CD3 zeta | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA | |
| A synthesized peptide derived from Human CD3 zeta [UniProt P20963] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 21H6 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 919 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title