missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Rat, Qfluor 630, Clone: 17A2, enQuireBio™
Rat Monoclonal Antibody
£204.75 - £274.05
Specifications
| Antigen | CD3 |
|---|---|
| Clone | 17A2 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15911808
|
EnquireBio
QAB3-QF630-100ug |
100 μg |
£204.75
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15921808
|
EnquireBio
QAB3-QF630-500ug |
500 μg |
£274.05
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD3 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 12502 | |
| IgG2b κ | |
| Protein A/G Purified |
| 17A2 | |
| Flow Cytometry | |
| Qfluor 630 | |
| RUO | |
| NP_033980 | |
| Cd3g | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title