Learn More
CD3 Mouse anti-Human, PE-Cy5, Clone: HIT3A, BD
Mouse Monoclonal Antibody
£57.73 - £226.63
Specifications
| Antigen | CD3 |
|---|---|
| Clone | HIT3A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE-Cyanine5 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15860679
|
BD Biosciences
561006 |
25 Tests |
£59.95 £57.73 / Each Save £2.22 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15858078
|
BD Biosciences
555341 |
100 Tests |
£236.00 £226.63 / Each Save £9.37 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HIT3a monoclonal antibody specifically binds to the human CD3 ε-chain, a 20 kDa subunit of the CD3/T cell antigen receptor complex found on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody can be mitogenic for T lymphocytes. The CD3 complex plays a role in signal transduction during antigen recognition by the T cell receptor for antigen. HIT3a antibody does not stain intracellular CD3 unlike the other CD3-specific clone, UCHT1.
Specifications
| CD3 | |
| Flow Cytometry | |
| PE-Cyanine5 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| HIT3A | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.