missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, APC, Clone: UCHT1, BD

Mouse Monoclonal Antibody

£58.53 - £603.80

Specifications

Antigen CD3
Clone UCHT1
Applications Flow Cytometry
Classification Monoclonal
Conjugate APC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15851359
View Documents
BD Biosciences
561810
25 Tests
ONLINE EXCLUSIVE PRICE
£61.00
£58.53 / Each
Save £2.47
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10106503
View Documents
BD Biosciences
555335
100 Tests
ONLINE EXCLUSIVE PRICE
£251.00
£240.34 / Each
Save £10.66
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15861359
View Documents
BD Biosciences
561811
500 Tests
ONLINE EXCLUSIVE PRICE
£1070.00
£603.80 / Each
Save £466.20
43% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The UCHT1 monoclonal antibody specifically binds to the human CD3ε-chain, a 20-kDa subunit of the CD3/T cell antigen receptor complex. CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody is mitogenic for CD3ε-positive cells when used in conjunction with costimulatory agents such as pokeweed mitogen or anti-CD28 antibody. CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε.

Specifications

Specifications

CD3
Flow Cytometry
APC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
UCHT1
Monoclonal
Mouse
Human
Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.