missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Canine, Clone: CA17.2A12, Novus Biologicals™
Mouse Monoclonal Antibody
£188.00 - £529.00
Specifications
| Antigen | CD3 |
|---|---|
| Clone | CA17.2A12 |
| Dilution | Flow Cytometry 1:25 - 1:50, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18139028
|
Novus Biologicals
NBP2-49891-0.025MG |
0.025 mg |
£188.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18617536
|
Novus Biologicals
NBP2-49891-0.1mg |
0.1 mg |
£529.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 Monoclonal specifically detects CD3 in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
| CD3 | |
| Flow Cytometry 1:25 - 1:50, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Canine | |
| CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Affinity enriched TCR/CD3 membrane proteins isolated from thymocytes and the T cell line CLGL-90 | |
| Primary |
| CA17.2A12 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| PBS | |
| 916 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title