missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Monoclonal Antibody (7D6), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD03014
Code : 50
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry.Specifications
CD3, CD3 | |
Monoclonal | |
FITC | |
CD3E | |
4930549J05Rik; A430104F18Rik; AI504783; antigen CD3D, delta polypeptide (TiT3 complex); antigen CD3E, epsilon polypeptide (TiT3 complex); antigen CD3G, gamma polypeptide; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; CD3; CD3 antigen delta chain; CD3 antigen delta polypeptide; CD3 antigen gamma chain; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; CD3 epsilon chain; CD3 epsilon subunit; CD3 epsilon subunit precursor; CD3 gamma-chain; CD3 glycoprotein; CD3 glycoprotein precursor; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3 protein; CD3 TCR complex; CD3 type I transmembrane glycoprotein; CD3 type I transmembrane glycoprotein precursor; CD3 zeta chain; Cd3d; CD3D antigen d | |
Mouse | |
purified | |
RUO | |
915, 916, 917, 919 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
7D6 | |
PBS with BSA and 0.1% sodium azide | |
P04234, P07766, P09693, P20963 | |
Cd247, Cd3d, CD3E, CD3g | |
Human CD3 epsilon | |
2 mL | |
Primary | |
Human | |
Antibody | |
IgG2a |