missing translation for 'onlineSavingsMsg'
Learn More

CD3 gamma Rabbit anti-Human, Rat, Clone: 7J2L5, Novus Biologicals™

Product Code. 18390843 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390843 100 μg 100µL
18367345 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18390843 Supplier Novus Biologicals Supplier No. NBP316327100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD3 gamma Monoclonal antibody specifically detects CD3 gamma in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3 gamma
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 7J2L5
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ79544, FLJ94613, MGC138597, T3G, T-cell antigen receptor complex, gamma subunit of T3, T-cell receptor T3 gamma chain, T-cell surface glycoprotein CD3 gamma chain
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 83-182 of human CD3 gamma (P09693).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 917
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.