missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD3 Fab-TACS™ Gravity Kit, Mouse
For manual positive cell separation. The Fab-TACS™ Gravity Kit provides label-free target cells without the use of high affinity antibodies and magnetic beads
£336.05 - £773.43
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Type | Non-magnetic Cell Isolation Kit |
Species | Mouse |
Sufficient For | 1x108 target cells/column |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16265350
|
IBA Lifesciences
6-3404-004 |
4 Columns |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16275350
|
IBA Lifesciences
6-3404-010 |
10 Columns |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Specifications
2°C to 8°C | |
Non-magnetic Cell Isolation Kit | |
Mouse | |
1x108 target cells/column |