missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunofluorescence, Functional Assay
Specifications
Specifications
| Antigen | CD3 epsilon |
| Applications | Flow Cytometry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Clone | RIV9 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunocytochemistry/Immunofluorescence, Functional |
| Formulation | 10 mM PBS |
| Gene Alias | CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?