missing translation for 'onlineSavingsMsg'
Learn More

CD3 epsilon Antibody (PC3/188A), DyLight 680, Novus Biologicals™

Product Code. 18424263 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424263 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424263 Supplier Novus Biologicals Supplier No. NBP254405FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Human, Rat, Porcine, Bovine, Chicken, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Negative), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3 epsilon
Applications Western Blot, Flow Cytometry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone PC3/188A
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 156-168 of the cytoplasmic domain of human CD3- chain (Uniprot: P07766 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 916
Target Species Human, Rat, Pig, Bovine, Chicken
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.