missing translation for 'onlineSavingsMsg'
Learn More

CD3 epsilon Antibody (OKT-3 (muromonab)), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18378210 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378210 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378210 Supplier Novus Biologicals Supplier No. NBP281031AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3 epsilon
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OKT-3 (muromonab)
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Gene Symbols CD3E
Host Species Rabbit
Immunogen Human peripheral E+ cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 916
Test Specificity Epitope on the epsilon-subunit within the human CD3 complex.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.