missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 epsilon Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£240.00 - £435.00
Specifications
| Antigen | CD3 epsilon |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18170199
|
Novus Biologicals
NBP2-38520 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600376
|
Novus Biologicals
NBP2-38520-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 epsilon Polyclonal specifically detects CD3 epsilon in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD3 epsilon | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| CD3E | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P07766 | |
| 916 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: WSKNRKAKAKPVTRGAGAGGRQRGQNKERPPPVPNPDYEPIRKGQRDLYSGLNQRRI | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title