missing translation for 'onlineSavingsMsg'
Learn More

CD3ε (Early T-Cell Marker) Ab-1 Rabbit Polyclonal Antibody, Epredia™

Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD3ε (Early T-Cell Marker) Ab-1, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-360-A1

 View more versions of this product

Product Code. 12663237

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD3 consists of five different polypeptide chains (designated as γ, δ, ε, ζ, and η). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition. CD3 antigen is a highly specific marker for T cells, and is present in majority of T cell neoplasms.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: aa 156-168 of ε-chain

Immunogen: A synthetic 13-mer peptide, corresponding to aa 156-168 of the ε-chain of human CD3 protein

Molecular Weight: ∽20kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD3e
Polyclonal
Rabbit
500 μL
T-Cell
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
A synthetic 13-mer peptide, corresponding to aa 156-168 of the epsilon-chain of human CD3 protein
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.