Learn More
CD3ε (Early T-Cell Marker) Ab-1 Rabbit Polyclonal Antibody, Epredia™
Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD3ε (Early T-Cell Marker) Ab-1, Rabbit Polyclonal Antibody.
Brand: Epredia RB-360-A
Description
CD3 consists of five different polypeptide chains (designated as γ, δ, ε, ζ, and η). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition. CD3 antigen is a highly specific marker for T cells, and is present in majority of T cell neoplasms.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: aa 156-168 of ε-chain
Immunogen: A synthetic 13-mer peptide, corresponding to aa 156-168 of the ε-chain of human CD3 protein
Molecular Weight: ∽20kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD3e | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| 1 mL | |
| T-Cell | |
| Human | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| 1:40-1:80 for 30 min RT | |
| A synthetic 13-mer peptide, corresponding to aa 156-168 of the epsilon-chain of human CD3 protein | |
| IVD | |
| Primary | |
| Store vial at 4deg.C. When stored at 2 to 8deg.C, | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.