missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 delta Rabbit anti-Human, Clone: 6, Novus Biologicals™
Rabbit Monoclonal Antibody
£308.00 - £448.00
Specifications
| Antigen | CD3 delta |
|---|---|
| Clone | 6 |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 ul/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217647
|
Novus Biologicals
NBP2-89729-50UL |
50 μg |
£308.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18233746
|
Novus Biologicals
NBP2-89729-100UL |
100 μg |
£448.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 delta Monoclonal specifically detects CD3 delta in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| CD3 delta | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 ul/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain | |
| Recombinant Human CD3 delta protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 6 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 915 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title