missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD3 delta/epsilon Antibody (203) [PerCP], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26882PCP
This item is not returnable.
View return policy
Description
CD3 delta/epsilon Monoclonal antibody specifically detects CD3 delta/epsilon in Mouse samples. It is validated for ELISA
Specifications
| CD3 delta/epsilon | |
| Monoclonal | |
| PerCP | |
| CD3 antigen, delta subunit, CD3 delta, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, IMD18, IMD19, OKT3, delta chain, T3D, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD3 delta/epsilon Heterodimer (Uniprot#: P04235 & P22646; Met1-Ala105 & Met1-Asp108). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 203 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 915 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction