missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 delta Antibody (TS-1), Novus Biologicals™
Mouse Monoclonal Antibody
£169.00
Specifications
| Antigen | CD3 delta |
|---|---|
| Clone | TS-1 |
| Concentration | 0.15 mg/mL |
| Dilution | Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay |
Description
CD3 delta Monoclonal specifically detects CD3 delta in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| CD3 delta | |
| 0.15 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction | |
| P04234 | |
| 915 | |
| Human TCR Vdelta1-Jdelta1 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| TS-1 | |
| Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain | |
| CD3D | |
| IgG1 | |
| Immunogen affinity purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title