missing translation for 'onlineSavingsMsg'
Learn More

CD3 delta Antibody, DyLight 550, Novus Biologicals™

Product Code. 18346313 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346313 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346313 Supplier Novus Biologicals Supplier No. NBP299673R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD3 delta Polyclonal specifically detects CD3 delta in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3 delta
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CD3 delta (NP_000723.1; Met 1-Ala 105).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 915
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.