missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD3+ CD45RO+ Tcm Fab Streptamer™ Isolation Kit MB
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- Strep-Tactin™ Magnetic Microbeads for Fab Streptamers™; 4 vials for 5 x 108 cells each
- CD3 Fab-Strep, human for isolation of T-cells; 1 vial for 1 x 109 cells
- CD45RO Fab-Strep, human for isolation of T-cells; 3 vials for 2 x 108 cells
- Biotin stock solution for Streptamer™ technology; 3 x 1 mL
- Buffer IS, Streptamer™ Washing Buffer, 10x concentrated; 2 x 50 mL
- Isolation of human CD3+ CD45RO+ central memory T cells is performed by serial double positive selection.
- In a first step PBMCs are labeled with magnetic CD3 Fab Streptamers™.
- Labeled cells are isolated in a strong magnet (StrepMan, horizontal position) where they migrate toward the tube wall on the side of the magnet.
- This CD3 positive cell fraction is collected and cells are liberated from all labeling reagents by addition of biotin in a strong magnet (StrepMan, upright position).
- The magnetic Streptamers™ migrate toward the tube wall and the label-free cells remain in the supernatant.
- Biotin is removed by washing.The pure and label-free CD3 positive cells are subjected to a second positive selection with magnetic CD45RO Fab Streptamers™ following the same procedure.
- The resulting cell preparation is highly enriched with CD3+ CD45RO+ central memory T cells with a purity of more than 90%.
- No columns are needed.
Specifications
Specifications
| Cell Type | PBMCs |
| Material | Magnetic Microspheres Coated with Strep-Tactin |
| No. of Cells | 1.6 x 109 |
| Quantity | 1 Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction