missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Antibody (OKT3) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NBP2-24867SS
This item is not returnable.
View return policy
Description
CD3 Monoclonal specifically detects CD3 in Human, Mouse samples. It is validated for Flow Cytometry, Functional, Immunoblotting, In vitro assay, In vivo assay, Flow (Cell Surface), CyTOF-ready.
Specifications
| CD3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1 uL/ 1 million cells, Functional, Immunoblotting reported in scientific literature (PMID 23880762), In vitro assay reported in scientific literature (PMID 26084721), In vivo assay reported in scientific literature (PMID 25535359), Flow (Cell Surface), CyTOF-ready | |
| anti-CD3 okt3, CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, okt3, okt3 ab, okt3 CD3, okt3 hybridoma, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Mouse | |
| 23.1 kDa | |
| 0.025 mg | |
| Adaptive Immunity, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells | |
| 916 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Functional Assay, Immunoblot, In vitro Assay, CyTOF | |
| OKT3 | |
| Unconjugated | |
| P04234 | |
| CD3 | |
| Human PBMCs were used as immunogen. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Functional activation was utilized in the following publication: An Epstein-Barr Virus (EBV) mutant with enhanced BZLF1 expression causes lymphomas with abortive lytic EBV infection in a humanized mouse model. Ma D S, X Yu, J E Mertz, J E Gumperz, E Reinheim, Y Zhou, W Tang, W J Burlingham, M L Gulley, S C Kenney. J Virol 86 7976-7987 (2012). This antibody is reactive to Human. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction