missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Antibody (CA17.2A12) [Allophycocyanin/Cy7], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-49891APCCY7
This item is not returnable.
View return policy
Description
CD3 Monoclonal antibody specifically detects CD3 in Canine samples. It is validated for Flow Cytometry
Specifications
| CD3 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Canine | |
| Purified |
| Flow Cytometry | |
| CA17.2A12 | |
| Flow Cytometry | |
| CD_antigen: CD3e, CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, IMD18, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Affinity enriched TCR/CD3 membrane proteins isolated from thymocytes and the T cell line CLGL-90 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells | |
| 916 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction