missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2F-10/SLAMF9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20382
This item is not returnable.
View return policy
Description
CD2F-10/SLAMF9 Polyclonal specifically detects CD2F-10/SLAMF9 in Human samples. It is validated for Western Blot.
Specifications
| CD2F-10/SLAMF9 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| SLAMF9 | |
| Recombinant protein encompassing a sequence within the center region of human SLAMF9. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 89886 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.85mg/mL | |
| Western Blot 1:500-1:3000 | |
| CD2 family member 10, CD2F10, CD2F-10MGC88194, CD84 homolog 1, CD84H1, CD84-H1cluster of differentiation 2 antigen family member 10, cluster of differentiation 84 homolog 1, SF2001, signaling lymphocytic activation molecule family member 2001, signaling lymphocytic activation molecule family member 9, SLAM family member 9 | |
| Rabbit | |
| 32 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction