missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD2BP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30698
This item is not returnable.
View return policy
Description
CD2BP2 Polyclonal antibody specifically detects CD2BP2 in Human,Mouse samples. It is validated for Western Blot
Specifications
| CD2BP2 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| CD2 (cytoplasmic tail) binding protein 2, CD2 antigen (cytoplasmic tail) binding protein 2, CD2 antigen (cytoplasmic tail)-binding protein 2, CD2 antigen cytoplasmic tail-binding protein 2, CD2 binding protein 2, CD2 cytoplasmic domain binding protein 2, CD2 cytoplasmic domain-binding protein 2, CD2 tail-binding protein 2, FWP010, KIAA1178, LIN1, Snu40, U5 snRNP 52K protein, U5-52K | |
| Between 275 and 325 | |
| 100 μg | |
| Immunology | |
| 10421 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction