missing translation for 'onlineSavingsMsg'
Learn More

CD2BP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £484.00

Specifications

Antigen CD2BP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18187397
View Documents
Novus Biologicals
NBP2-47516
0.1 mL
£484.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18637375
View Documents
Novus Biologicals
NBP2-47516-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD2BP2 Polyclonal specifically detects CD2BP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CD2BP2
Polyclonal
Rabbit
Immunology
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
10421
This antibody was developed against a recombinant protein corresponding to amino acids: LGPHNPTPPPSLDMFAEELAEEELETPTPTQRGEAESRGDGLVDVMWEYKWENTGDAELYGPFTSAQMQTWVSEGYFPDGVYCRKLDPPGGQFYNSKRIDFDLY
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
CD2 (cytoplasmic tail) binding protein 2, CD2 antigen (cytoplasmic tail) binding protein 2, CD2 antigen (cytoplasmic tail)-binding protein 2, CD2 antigen cytoplasmic tail-binding protein 2, CD2 binding protein 2, CD2 cytoplasmic domain binding protein 2, CD2 cytoplasmic domain-binding protein 2, CD2 tail-binding protein 2, FWP010, KIAA1178, LIN1, Snu40, U5 snRNP 52K protein, U5-52K
CD2BP2
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.