missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: WSGTLNNKLG LFPSNFVKEL EVTDDGETHE AQDDSETVLA GPTSPIPSLG NVSETASGSV TQPKKIRGIG FGDIFKEGSV KLRTRTSSSE TEEKKPEKPL ILQSLGPKTQ SVEITKTDTE GKIKAKE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
This gene encodes a scaffolding molecule that regulates the actin cytoskeleton. The protein directly interacts with filamentous actin and a variety of cell membrane proteins through multiple actin binding sites, SH3 domains, and a proline-rich region containing binding sites for SH3 domains. The cytoplasmic protein localizes to membrane ruffles, lipid rafts, and the leading edges of cells. It is implicated in dynamic actin remodeling and membrane trafficking that occurs during receptor endocytosis and cytokinesis. Haploinsufficiency of this gene is implicated in susceptibility to glomerular disease.
Specifications
Specifications
| Antigen | CD2AP |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CD2AP |
| Gene Accession No. | F1LRS8, Q9JLQ0, Q9Y5K6 |
| Gene Alias | adapter protein CMS; AL024079; C78928; Cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2 associated protein; Cd2ap; CD2-associated protein; CMS; mesenchyme to epithelium transition protein, SH3 domains; mesenchyme-to-epithelium transition protein with SH3 domains 1; Mets1; METS-1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1 |
| Gene Symbols | CD2AP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
