missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD299 (DC-SIGN/L) Monoclonal Antibody (2G1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521012
This item is not returnable.
View return policy
Description
Peptide Sequence: SQRNWHDSVT ACQEVRAQLV VIKTAEEQNF LQLQTSRSNR FSWMGLSDLN QEGTWQWVDG SPLSPSFQRY WNSGEPNNSG NEDCAEFSGS GWNDNRCDVD NYWICKKPAA.
CLEC4M is a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells.Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.This gene encodes a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells. This gene is mapped to 19p13.3, in a cluster with the CD209 and CD23/FCER2 genes. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined.
Specifications
| CD299 (DC-SIGN/L) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9H2X3 | |
| CLEC4M | |
| CLEC4M (NP_055072, 285 a.a. ~ 394 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 λ |
| ELISA, Western Blot | |
| 2G1 | |
| Unconjugated | |
| CLEC4M | |
| CD209 antigen-like protein 1; Cd209b; CD209b antigen; CD209L; CD209L1; CD299; CD299 antigen; CLEC4M; C-type lectin domain family 4 member M; C-type lectin domain family 4, member M; DC-SIGN2; DCSIGNR; DC-SIGNR; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin 2; HP10347; liver/lymph node-specific ICAM-3 grabbing non-integrin; liver/lymph node-specific ICAM-3-grabbing non-integrin; LSIGN; L-SIGN; mannose binding C-type lectin DC-SIGNR; MGC129964; MGC47866 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10332 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction