missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD298 Recombinant Rabbit Monoclonal Antibody (23GB4740), Invitrogen™
GREENER_CHOICE

Product Code. p-200088066
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089898 100 μL 100µL
30089646 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089898 Supplier Invitrogen™ Supplier No. MA553802

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 3 subunit. This gene encodes a beta 3 subunit. A pseudogene exists for this gene, and it is located on chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD298
Applications Western Blot
Classification Recombinant Monoclonal
Clone 23GB4740
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATP1B3
Gene Accession No. P54709, P97370, Q63377
Gene Alias AA409958; AI664000; Atp1b3; ATPase Na+/K+ transporting subunit beta 3; ATPase, Na+/K+ beta 3 polypeptide; ATPase, Na+/K+ transporting, beta 3 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 3; ATPB-3; AW212096; CD298; CD298 antigen; Na K-ATPase beta-3 subunit; Na+/K+ -ATPase beta 3 subunit; NKAB3S; Sodium Potassium ATPase; Sodium/potassium-dependent ATPase subunit beta-3; sodium/potassium-transporting ATPase subunit beta-3
Gene Symbols ATP1B3
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATP1B3 (241-279AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11933, 25390, 483
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.